Learn More
TPP™ Centrifuge Tubes
Intended for centrifugation of suspensions such as cells, bacteria, precipitate and others. TPP™ Centrifuge Tubes are manufactured on automatic transport units ensuring quick and careful handling without unnecessary contamination risk.
Brand: TPP™ 91115
Description
Intended for centrifugation of suspensions such as cells, bacteria, precipitate and others. TPP™ Centrifuge Tubes are manufactured on automatic transport units ensuring quick and careful handling without unnecessary contamination risk.
- Manufactured with highest grade of raw material fulfilling requirements of USP Class VI
- White writing circle on screw cap for easy labeling
- Instrument caps cannot be over-tightened; gas and aerosol tight cap
- Graduations down to 0.1 ml / 0.5 ml respectively
- Convenient autoclavable, collapsible racks available
Specifications
15 mL | |
Polystyrene | |
Conical | |
Yes |
Single-use | |
120 mm | |
17.1 mm | |
1700 x g |