Learn More
CD86 Rat anti-Mouse, FITC, Clone: GL1, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech FITC-65068-500UG
Additional Details : Weight : 0.00010kg
Description
The CD86 antibody from Proteintech is a rat monoclonal antibody generated with lps-activated cba/ca mouse spleen b cells of mouse CD86. This antibody recognizes mouse antigen. The CD86 antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
CD86 | |
Monoclonal | |
0.5 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
GL1 | |
FITC | |
500 μg | |
12524 | |
IgG2a |