Learn More
Calcium shot, 99.5% (metals basis), Thermo Scientific™ Product Code.: 11341178
Calcium shot, 99.5% (metals basis), Thermo Scientific™
Product Code. 11341178
2 x 500 g
Click to view available options
Quantity:
5 g
50 g
250 g
2 x 500 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11341178
Brand: Thermo Scientific Alfa Aesar 010127.95
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]
Specifications
Color Silver-White
Appearance Silvery-white shot
Assay Percent Range 99.5% (Metals basis)
Quantity 2 x 500 g
Odor Odorless
Vapor Pressure 0 hPa
Form Shot
Chemical Name or Material Calcium
Product Content Correction
Product Title
Calcium shot, 99.5% (metals basis), Thermo Scientific™ >