Learn More
Calcium chloride, Free-flowing, anhydrous, ≥97%, Honeywell Fluka™
Product Code. 15677020
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Free-flowing, anhydrous,≥97%
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]
Specifications
Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Quantity 100 g
Linear Formula CaCl2
IUPAC Name calcium dichloride
Formula Weight 110.98
Percent Purity >97%
Packaging Poly Bottle
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Free-flowing, anhydrous, ≥97%