missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, Free-flowing, anhydrous, ≥97%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Boiling Point | 1,900°C |
Melting Point | 772°C (literature) |
CAS Max % | 100.0000% |
Quantity | 500 g |
Linear Formula | CaCl2 |
IUPAC Name | calcium;dichloride |
Formula Weight | 110.98 |
Percent Purity | >97% |
Packaging | Poly Bottle |
Density | 2.150 g/cm3 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction