Learn More
Epredia™ Cabinet Colorstack Slide
Brand:  Epredia™ E43/12C/GREEN
Additional Details : Weight : 12.00000kg
Product Code. 12161820
  • 374.00 EUR / Each
Estimated Shipment: 12-08-2024
Description
14 drawer holds 76mm x 26mm microscope slides vertically 6000 slide capacity green Epredia™ Raymond Lamb
Specifications
Specifications
Cabinets with Drawers
6000 Slides
14 cm
Green
Slide storage
48 cm