Learn More
Macherey-Nagel™ Butyl Phase Polygosil™ C4
218.00 EUR - 1905.00 EUR
Specifications
Particle Size | 7 μm |
---|---|
Technique | Liquid Chromatography |
Type | Polygosil C4 Phases |
Product Code | Brand | Quantity | Particle Size | Pore Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Particle Size | Pore Size | Price | Quantity & Availability | |||
10505781
|
Macherey-Nagel™
711991.100 |
100 g | 7 μm | 1000 Å |
1905.00 EUR
100g |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10566291
|
Macherey-Nagel™
711680.100 |
100 g | 7 μm | 300 Å |
1905.00 EUR
100g |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10661482
|
Macherey-Nagel™
711991.10 |
10 g | 7 μm | 1000 Å |
218.00 EUR
10g |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10556291
|
Macherey-Nagel™
711680.10 |
10 g | 7 μm | 300 Å |
218.00 EUR
10g |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
Specifications
7 μm | |
Liquid Chromatography | |
Polygosil C4 Phases |