Learn More
IKA™ BMT-20-S Tubes
Brand: IKA™ 0003703200
258.32 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
BMT-20-S Tube | |
Dry milling of dry and brittle samples (e.g. kaolin, gypsum, colored pigments, tablets), cell maceration, processing of materials mixed with fluids | |
25 Pk. |
2 to 15 mL | |
BMT-20-S |
Special Offers
- Multiple Manufacturers
Product Content Correction
Product Title
IKA™ BMT-20-S Tubes > Capacity: 2 to 15mL
Spot an opportunity for improvement?Share a Content Correction