Learn More
IKA™ BMT-20-G Tubes
Brand:  IKA™ 0003703300
258.32 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10263035
  • 278.00 EUR / Pack of 25
In Stock
Specifications
Tube
Dry milling of dry and brittle samples (e.g. kaolin, gypsum, colored pigments, tablets), cell maceration, processing of materials mixed with fluids
25 Pk.
2 to 15 mL
BMT-20-G
Documents
One moment while we fetch your results.
Product Title
IKA™ BMT-20-G Tubes > Capacity: 2 to 15mL