Learn More
Grant Instruments™ Blanket for Inspissator
Provides thermal insulation and minimizes drafts. Grant Instruments™ Blanket for Inspissator is insect resistant for use with production of tuberculosis culture medium
Brand: Grant Instruments™ BIN
Additional Details : Weight : 2.00000kg
Description
- For use with T100IN Inspissator
- Insect resistant
- Provides thermal insulation and minimizes drafts when placed over the containers
Specifications
Inspissator |