Learn More
Black Eriochrome T, Chem Lab™
Click to view available options
Concentration:
10 g/L
5 g/L
Specifications
Specifications
Concentration | 5 g/L |
Solubility Information | Soluble in water |
Odor | Specific |
Color | Black |
Quantity | 1 L |
Physical Form | Liquid |