missing translation for 'onlineSavingsMsg'
Learn More

Bis(triphenylphosphine)copper(I) borohydride, Thermo Scientific™

Product Code. 11431940
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16903-61-0
Molecular Formula C36H34BCuP2
Molecular Weight (g/mol) 602.97
MDL Number MFCD00013219
InChI Key YVXRFTLDBYUBGJ-UHFFFAOYSA-N
Synonym bis triphenylphosphine copper tetrahydroborate, bis triphenylphosphine copper i borohydride, bis-triphenylphosphino-cuprous borohydride, tetrahydridoboratobis triphenylphosphine copper i, bis triphenylphosphine copper i tetrahydridoborate, bis triphenylphosphine copper tetrahydroborate,? pph3 2cubh4
PubChem CID 10908163
IUPAC Name boron(1-);copper(1+);triphenylphosphane
SMILES [BH4-].[Cu+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼175°C (decomposition)
Linear Formula [(C6H5)3P]2CuBH4
Quantity 25 g
UN Number UN2813
Sensitivity Moisture sensitive
Formula Weight 602.97
Chemical Name or Material Bis(triphenylphosphine)copper(I) borohydride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.