Learn More
Lonza™ BioWhittaker™ Hank's Balanced Salt Solutions (1X)
Used in wide variety of tissue culture applications. BioWhittaker™ Hank's Balanced Salt Solutions (1X) is used to maintain a physiological pH for cells maintained in non-CO2 conditions, diluting cells before counting, washing cells before dissociating and transporting cells or tissues.
22.65 EUR - 131.00 EUR
Specifications
Content And Storage | 15° to 30°C |
---|---|
Product Type | Hanks' Balanced Salt Solution |
Sterility | Sterile |
Product Code | Brand | Formulation | Quantity | pH | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | pH | Price | Quantity & Availability | |||
11625160
|
Lonza™
10-543Q |
1 L | 7.14 to 7.46 | N/A | |||||
11675150
|
Lonza™
10-508Q |
1 L | 7.1 to 7.4 | N/A | |||||
11615160
|
Lonza™
BE10-543F |
500 mL | 7.14 to 7.46 | N/A | |||||
11635160
|
Lonza™
BE10-547F |
500 mL | 7.14 to 7.46 | N/A | |||||
11685150
|
Lonza™
BE10-527F |
500 mL | 7.08 to 7.4 | N/A | |||||
11655120
|
Lonza™
04-315Q |
1 L | 7.14 to 7.46 | N/A | |||||
11665150
|
Lonza™
BE10-508F |
500 mL | 7.1 to 7.4 | N/A | |||||
Description
- Available in various formulations including calcium-, magnesium- and phenol-red-free or containing versions
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
- Contain NaHCO3
Order Info
4L solution is packaged in a polybag.
Specifications
15° to 30°C | |
Hanks' Balanced Salt Solution | |
Sterile |