Learn More
Erlab™ Bench Cabinet with Carbon Filtration
Erlab™ Bench Cabinet with Carbon Filtration is designed to reduce the inhalation risks associated with the concentration of vapors into the lab environment.
Brand: Erlab™ B01IA023/BE
Description
- Non-ducted storage cabinet
- High level of protection
- Mobile
- Flexible
- Environmentally friendly
- Low energy consumption
- Elimination of harmful, odoros vapors
- Quiet ventilation system
- High corrosion resistance
- Lockable
- Flat packed to facilitate laboratory access
- Designed to store a wide variety of reagents used in the laboratory
Specifications
BE (Polyvalent for Acids and Organic Vapors) | |
Storage Cabinet |