Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
Brand:  BD 309628
 View more versions of this product
Product Code. 15880305
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
  • Sterile syringe for single use
  • Without needle
  • Clear barrel (polycarbonate or polypropylene) with bold scale markings
  • Tapered plunger rod for ease of aspiration
  • Positive plunger rod stop
  • Added BD Luer-Lok™ thread for increased secure connection
  • DNR (Latex) free
  • Shelf life: 1825 days
Specifications
Specifications
Syringe
No Needle Included
Luer Lock
Single-use
1 mL
Sterile
Product Title
BD Disposable Syringes with Luer-Lok™ Tips > Luer-Lok; Volume: 1mL