Learn More
SCAT Base for space saving canister
Click to view available options
Material: Stainless steel, Typ Canister Holder
Description
- Made of stainless steel
- Base for 2 space saving containers
SCAT space-saving canisters offer a clever alternative to conventional SCAT canisters by requiring only a fraction of the floor space for storage, without compromising on capacity
Specifications
Specifications
Description | Base for 2 space saving containers |
Dimensions (L x W x H) | 200 x 141 x 130 mm |
Material | Stainless Steel |