Learn More
Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack
Specifically designed for eliminating sample retention within the pipette tip.
53.00 EUR - 86.60 EUR
Specifications
Filtered Non-Filtered
Color Clear
Product Line ART
View More Specs
Products 6
Product Code Brand Description Volume (Metric) Tip Style Sterility Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Tip Style Sterility Price Quantity & Availability  
10110062
View Documents Promotion Details
Thermo Scientific™
3771-05
ART 300, Non-Filtered, Sterile, Rack 300 μL MicroPoint Sterile
86.60 EUR
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
10550682
View Documents Promotion Details
Thermo Scientific™
3772-05
ART 300, Non-Filtered, Non-sterile, Rack 300 μL MicroPoint Non-sterile
78.00 EUR
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
10261722
View Documents Promotion Details
Thermo Scientific™
3511-05
ART 10 REACH, Non-filtered, Sterile, Racked, Lift-off Lid 10 μL Low Retention Sterile
70.40 EUR
Pack of 960
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
10251722
View Documents Promotion Details
Thermo Scientific™
3512-05
ART 10 REACH, Non-filtered, Non-sterile, Racked, Lift-off Lid 10 μL Low Retention Non-sterile
56.00 EUR
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
10241772
View Documents Promotion Details
Thermo Scientific™
3521-05
ART 20, Low Retention, Non-filtered, Sterile, Racked, Lift-off Lid 20 μL Low Retention Sterile
73.00 EUR
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
10048481
View Documents Promotion Details
Thermo Scientific™
3522-05
ART 20 Low Retention, Non-filtered, Non-sterile, Racked, Lift-off Lid 20 μL Low Retention Non-sterile
53.00 EUR
Pack of 960
View alternative products
Save Up to 
»
Estimated Shipment: 06-09-2024
Description
Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack are specifically designed for eliminating sample retention within the pipette tip.
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Tips seal when exposed to potential contaminants
  • DNase, RNase, DNA, ATP, and Endotoxin Free
Spezifikation
Spezifikation
Non-Filtered
Clear
ART
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Product Certifications