missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Persulfate, ACS, ≥98%, Ultrapure

Product Code. 15895228 Shop All Thermo Scientific Products
100 g, Plastic bottle, 100g
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
10g
1kg
This item is not returnable. View return policy
This item is not returnable. View return policy

High speed initiator used to catalyze acrylamide polymerization. | CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol

A high speed initiator used to catalyze acrylamide polymerization.

Working Stock Solution (10%):

  • Ammonium Persulfate - 1 gm
  • dH2O to 10 ml
  • This solution should be stored at 4°C and prepared fresh weekly.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diazanium;sulfonatooxy sulfate
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS(=O)(=O)[O-]

Specifications

Melting Point 120°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
UN Number UN1444
Formula Weight 228.19
Percent Purity ≥98%
Grade ACS
Packaging Plastic bottle
Specific Gravity 1.982
Density 1.982
Chemical Name or Material Ammonium Persulfate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Persulfate, ACS, ≥98%, Ultrapure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.