missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium chloride, 0.5M aq. soln., Thermo Scientific™
Click to view available options
Quantity:
50 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Ammonium chloride is used for the lysis of human red blood cells. It is used in protein isolation techniques and in the study of basic calcium phosphate crystals in fibroblasts. It inhibits intracellular dissolution of basic calcium phosphate (BCP) crystals by alkalinizing phagolysosomes. It is used as a flavoring agent in some types of liquorices. It is also used as electrolyte in zinc-carbon batteries.
Solubility
Fully miscible in water. Soluble in ethanol, methanol, and glycerol. Insoluble in acetone, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Ammonium chloride is used for the lysis of human red blood cells. It is used in protein isolation techniques and in the study of basic calcium phosphate crystals in fibroblasts. It inhibits intracellular dissolution of basic calcium phosphate (BCP) crystals by alkalinizing phagolysosomes. It is used as a flavoring agent in some types of liquorices. It is also used as electrolyte in zinc-carbon batteries.
Solubility
Fully miscible in water. Soluble in ethanol, methanol, and glycerol. Insoluble in acetone, and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | NH4Cl |
Beilstein | 4371014 |
Solubility Information | Fully miscible in water. Soluble in ethanol,methanol,and glycerol. Insoluble in acetone,and ether. |
Formula Weight | 53.49 |
Concentration or Composition (by Analyte or Components) | 0.5M aq. soln. |
Chemical Name or Material | Ammonium chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction