Learn More
Aluminum oxide substrate, 10x10x0.5mm, polished one side, A plane, Thermo Scientific Chemicals
110.55 EUR - 462.30 EUR
Specifications
Chemical Name or Material | Aluminum Oxide Substrate, A Plane |
---|---|
Name Note | Polished One Side |
Molecular Formula | Al2O3 |
MDL Number | MFCD00003424 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308847
|
Thermo Scientific Alfa Aesar
038498.KT |
1 Ea. |
110.55 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398837
|
Thermo Scientific Alfa Aesar
038498.KU |
5 Ea. |
462.30 EUR
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum Oxide Substrate, A Plane | |
Polished One Side | |
Al2O3 | |
MFCD00003424 |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures