Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
Description
- Velcro fastening to neck
- Tie fastening at the waist
- Double cuff with knitted
- inner cuff
- Welded and taped seams
Specifications
Specifications
Type | Apron |
Size | 4X-Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Green |
Disposable | Limited-use |
Closure Type | Hook and Loop, Waist Ties |
Sterility | Non-sterile |
Cuff Style | Double Inner Knit |
Latex Free | Yes |
No. of Pockets | None |
Show More |
Product Content Correction
Product Title
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves >
Spot an opportunity for improvement?Share a Content Correction