Learn More
Al-23 Tube, One End Closed, O.D.: 6mm, I.D.: 3mm, Thermo Scientific™
27.90 EUR - 174.00 EUR
Specifications
Chemical Name or Material Al-23 Tube
Name Note One end closed
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362728
View Documents
Thermo Scientific Alfa Aesar
030064.DM
100 mm N/A N/A N/A
11352728
View Documents
Thermo Scientific Alfa Aesar
030064.DR
200 mm N/A N/A N/A
11332728
View Documents
Thermo Scientific Alfa Aesar
030064.DW
300 mm N/A N/A N/A
11372728
View Documents
Thermo Scientific Alfa Aesar
030064.EH
600 mm N/A N/A N/A
11382728
View Documents
Thermo Scientific Alfa Aesar
030064.EM
800 mm N/A N/A N/A
11392728
View Documents
Thermo Scientific Alfa Aesar
030064.EO
1000 mm N/A N/A N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube
One end closed
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Al-23 Tube, One End Closed, O.D.: 6mm, I.D.: 3mm, Thermo Scientific™