Learn More
Al-23 Tube, Both Ends Open, O.D.: 30mm, I.D.: 25mm, Thermo Scientific Chemicals
62.89 EUR - 485.00 EUR
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11334007
View Documents
Thermo Scientific Alfa Aesar
031574.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11394007
View Documents
Thermo Scientific Alfa Aesar
031574.DR
200 mm
123.62 EUR
200mm
11344007
View Documents
Thermo Scientific Alfa Aesar
031574.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354007
View Documents
Thermo Scientific Alfa Aesar
031574.EH
600 mm
362.81 EUR
600mm
11384007
View Documents
Thermo Scientific Alfa Aesar
031574.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11374007
View Documents
Thermo Scientific Alfa Aesar
031574.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures