Learn More
Brady™ Aggressive Adhesive Die-cut Multipurpose Polyester Labels for M6/M7 Printers
Thermal transfer printable, glossy permanent polyester labels are ideal for use in all commercial and industrial applications.
44.30 EUR - 282.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
Certifications/Compliance | ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized |
Adhesive Type | Permanent Acrylic |
Coating | None |
Color | White |
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (LxW) | For Use With (Equipment) | Height (Metric) | Shape | Price | Quantity & Availability | |||
17828114
|
Brady™
174333 |
0.2 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 5.08 mm | Rectangle |
73.55 EUR
Pack of 750 |
In Stock
Log in to see stock. |
|||
17858204
|
Brady™
174327 |
0.375 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
54.40 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17828974
|
Brady™
174329 |
0.375 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
76.70 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17888194
|
Brady™
174196 |
0.375 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 9.53 mm | Rectangle |
73.50 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17878224
|
Brady™
173320 |
0.375 x 1.5 in. | M710, BMP71 | 9.53 mm | Rectangle |
70.95 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17868284
|
Brady™
174344 |
0.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Square |
61.90 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17818164
|
Brady™
174206 |
0.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
73.50 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17838174
|
Brady™
174255 |
0.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 12.7 mm | Rectangle |
90.30 EUR
Pack of 500 |
In Stock
Log in to see stock. |
|||
17868324
|
Brady™
174304 |
0.6 x 0.375 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 15.24 mm | Rectangle |
89.95 EUR
Pack of 750 |
Estimated Shipment: 18-06-2025
Log in to see stock. |
|||
17809104
|
Brady™
173354 |
0.75 x 1.5 in. | M710, BMP71 | 19.05 mm | Rectangle |
73.15 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17839164
|
Brady™
174266 |
0.75 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 19.05 mm | Rectangle |
79.00 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17888184
|
Brady™
174219 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Square |
57.30 EUR
Pack of 250 |
In Stock
Log in to see stock. |
|||
17898724
|
Brady™
174455 |
1 x 2 in. | M610, M710, M611, BMP61, BMP71 | 25.4 mm | Rectangle |
282.00 EUR
Pack of 1000 |
In Stock
Log in to see stock. |
|||
17848434
|
Brady™
174239 |
1 x 3 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
50.00 EUR
Pack of 100 |
Estimated Shipment: 18-06-2025
Log in to see stock. |
|||
17811479
|
Brady™
173758 |
1 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 25.4 mm | Rectangle |
54.10 EUR
Pack of 100 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||
17871499
|
Brady™
173831 |
1.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 38.1 mm | Square |
83.20 EUR
Pack of 250 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||
17818334
|
Brady™
174291 |
1.9 x 3 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 48.26 mm | Rectangle |
65.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||
17889034
|
Brady™
173330 |
1.9 x 3 in. | M710, BMP71 | 48.26 mm | Rectangle |
60.25 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||
17808104
|
Brady™
174298 |
1.9 x 4 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 48.26 mm | Rectangle |
81.40 EUR
Pack of 100 |
Estimated Shipment: 18-06-2025
Log in to see stock. |
|||
17828774
|
Brady™
174227 |
2 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | 50.8 mm | Rectangle |
44.30 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
- Aggressive adhesive good for powder-coated and textured flat surfaces including low surface energy plastics
- Designed to withstand exposure to numerous solvents
- UL approved, CSA accepted
- Service temperature of -40°C to 100°C (-40°F to 212°F)
- Made of B-422 polyester material
Specifications
Not Autoclavable | |
Permanent Acrylic | |
White | |
Die-cut | |
Single-side Printable | |
Labels | |
Asset Tracking, Circuit Board Labeling, Equipment Labeling, General Identification | |
-40°F to +212°F | |
0.004 in. |
ASTM B 117, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized | |
None | |
Gloss | |
Polyester | |
Thermal Transfer | |
Non-sterile | |
No | |
-40°C to +100°C | |
0.102 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ Aggressive Adhesive Die-cut Multipurpose Polyester Labels for M6/M7 Printers