Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Fluorodyne™ II Membrane Optimized for Scale Up
Fluorodyne II membrane is a hydrophilic polyvinylidene fluoride (PVDF) that offers higher flow rates with low protein and preservative binding, and broad chemical and temperature resistance.
Brand:  Cytiva (formerly Pall Lab) 4907
Additional Details : Weight : 0.19000kg
Product Code. 17114451
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Higher flow rates with low protein and preservative binding
  • Broad chemical and temperature resistance
Specifications
Specifications
PVDF
0.2 μm