Learn More
Cytiva (formerly Pall Lab) Acrodisc™ PSF Syringe Filters with PTFE Membrane
The retention efficiency of the Acrodisc PSF filter can directly increase the life of your HPLC column.
166.00 EUR - 3230.00 EUR
Specifications
Sterility | Non-sterile |
---|---|
Filtration Area | 3.9 cm2 |
Housing Material | Polypropylene |
Diameter (Metric) | 25 mm |
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | No. per Pack | Material (Membrane) | Price | Quantity & Availability | |||
17164661
|
Cytiva (formerly Pall Lab)
AP-4219 |
0.45 μm | 50/Pk. | PTFE |
166.00 EUR
Pack of 50 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
17174661
|
Cytiva (formerly Pall Lab)
AP-4225 |
0.2 μm | 50/Pk. | PTFE |
166.00 EUR
Pack of 50 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||
17104681
|
Cytiva (formerly Pall Lab)
AP-4501 |
0.45 μm | 1000/Pk. | PTFE |
2890.00 EUR
Pack of 1000 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17124681
|
Cytiva (formerly Pall Lab)
AP-4521 |
0.2 μm | 1000/Pk. | PTFE |
2890.00 EUR
Pack of 1000 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
|||
17194661
|
Cytiva (formerly Pall Lab)
AP-4302 |
0.45 μm | 1000/Pk. | Glass Fiber, PTFE |
3230.00 EUR
Pack of 1000 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||
17154691
|
Cytiva (formerly Pall Lab)
AP-4790 |
0.2 μm | 50/Pk. | Glass Fiber, PTFE |
196.00 EUR
Pack of 50 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
17104671
|
Cytiva (formerly Pall Lab)
AP-4303 |
0.45 μm | 50/Pk. | Glass Fiber, PTFE |
183.00 EUR
Pack of 50 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17164691
|
Cytiva (formerly Pall Lab)
AP-4791 |
0.2 μm | 1000/Pk. | Glass Fiber, PTFE |
3190.00 EUR
Pack of 1000 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
17184661
|
Cytiva (formerly Pall Lab)
AP-4301 |
0.45 μm | 25/AutoPack Tubes | Glass Fiber, PTFE |
835.00 EUR
Pack of 200 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
Description
- More retentive filters remove smaller non-dissolved particles that may rest on the head of the HPLC column and affect analytical results as they dissolve during an analysis
Specifications
Non-sterile | |
3.9 cm2 | |
Polypropylene | |
25 mm |