Learn More
Acetone-d{6}, 100%(Isotopic), contains 0.03% v/v TMS, Thermo Scientific Chemicals
71.15 EUR - 287.43 EUR
Chemical Identifiers
CAS | 666-52-4 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 64.117 |
MDL Number | MFCD00044635 |
InChI Key | CSCPPACGZOOCGX-WFGJKAKNSA-N |
Synonym | acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone |
PubChem CID | 522220 |
ChEBI | CHEBI:78217 |
IUPAC Name | 1,1,1,3,3,3-hexadeuteriopropan-2-one |
SMILES | CC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359937
|
Thermo Scientific Alfa Aesar
042263.SA |
2 Ea. |
71.15 EUR
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349937
|
Thermo Scientific Alfa Aesar
042263.KV |
10 Ea. |
287.43 EUR
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
666-52-4 | |
64.117 | |
CSCPPACGZOOCGX-WFGJKAKNSA-N | |
522220 | |
1,1,1,3,3,3-hexadeuteriopropan-2-one |
C3H6O | |
MFCD00044635 | |
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone | |
CHEBI:78217 | |
CC(=O)C |
Specifications
Liquid | |
0.872 g/mL | |
Acetone-like | |
666-52-4 | |
56°C | |
MFCD00044635 | |
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone | |
CC(=O)C | |
64.117 | |
CHEBI:78217 | |
100% | |
Acetone-d6, Isotopic |
Colorless | |
-17°C (1°F) | |
1.3554 | |
-94°C | |
C3H6O | |
UN1090 | |
CSCPPACGZOOCGX-WFGJKAKNSA-N | |
1,1,1,3,3,3-hexadeuteriopropan-2-one | |
522220 | |
64.13 | |
contains 0.03% v/v TMS |
Safety and Handling
GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H225-H319-H335-H336-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE
missing translation for 'einecsNumber' : 211-563-9
missing translation for 'tsca' : No
Recommended Storage : Ambient
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.