missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acepromazine maleate, 98%

Catalog Number p-7214420
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
3598-37-6
C23H26N2O5S
442.53
FQRHOOHLUYHMGG-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O
This item is not returnable. View return policy
3598-37-6
C23H26N2O5S
442.53
FQRHOOHLUYHMGG-WLHGVMLRSA-N
(2E)-but-2-enedioic acid; 1-{10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl}ethan-1-one
OC(=O)\C=C\C(O)=O.CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 135.0°C to 138.0°C
Color Clear Colorless to Yellow
Assay Percent Range 97.5 % min. (HPLC)
Quantity 250 mg
Solubility Information Solubility aq. soln.: 37 g/L (20°C).
Formula Weight 442.53
Chemical Name or Material Acepromazine maleate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'