Learn More
Fisherbrand™ 96 PP Well Plate with 96 Glass Insert and PE Snap Caps with Septum
Description
- 96 Wells
- Silicone/PTFE
- Clear
- Glass
- Plates meet ANSI and SBS standards
- Different sealing options
- Chromatography tested
- No leeching due to high-end material
Specifications
Standard | |
Clear | |
96 | |
Taper |
8 x 12 | |
Glass | |
400 μL | |
Taper |
Product Content Correction
Product Title
Fisherbrand™ 96 PP Well Plate with 96 Glass Insert and PE Snap Caps with Septum > 9mm cap, Silicone/PTFE seal, Pre-slit, 45°,400μL,taper bottom
Spot an opportunity for improvement?Share a Content Correction