Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Shop All Cole Parmer Products
Click to view available options
Holds:
4 x 96-well PTFE Microplates
40 x 24 mm Tubes
96 x 16 mm Tubes
Description
- Compatible with Electrothermal RS9000 Reaction Stations
Specifications
Specifications
Description | Heating Block for Reaction System |
For Use With (Equipment) | RS-250D-S Shaking Reaction Stations |
Holds | 96 x 16 mm Tubes |
No. of Places | 96 |
Type | Heating Block |
Warranty | 1 Year |
Diameter (English) Outside | 0.6 in. |
Diameter (Metric) Outside | 16 mm |
Product Content Correction
Product Title
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations >
Spot an opportunity for improvement?Share a Content Correction