Learn More
Ansell™ 70-225 Series Yellow Neptune™ Heavy Weight Gloves
100% aramid fiber construction offers a very high cut resistance to weight ratio. Repels liquids, avoiding damage to the strands and extending glove life.
281.00 EUR
Specifications
Back Material | Neptune™ |
---|---|
Certifications/Compliance | Standards CATEGORY III EN 388 : 254X, EN 407, ANSI |
Cuff Material | Neptune™ |
Product Type | Heavyweight Gloves |
Glove Cuff Style | Knit |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12636921
|
Ansell™
70225100 |
10 |
281.00 EUR
12 pairs |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
12656921
|
Ansell™
70225080 |
8 |
281.00 EUR
12 pairs |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
12646921
|
Ansell™
70225070 |
7 |
281.00 EUR
12 pairs |
Estimated Shipment: 03-03-2025
Log in to see stock. |
|||||
Description
- Made from high-tech Kevlar™ aramid fibers para-aramid short fibers
- Neptune coating allows glove fibers to repel liquids
- Seamless construction to avoid irritation
- Short fibers provide extra comfort
- Terry-loop technique enchances heat insulation
- Thick wall offers high shock absorption
- Ambidextrous
- Washable and reusable
Specifications
Neptune™ | |
Neptune™ | |
Knit | |
Dupont™ Kevlar™ aramid fibers | |
Very high cut resistance |
Standards CATEGORY III EN 388 : 254X, EN 407, ANSI | |
Heavyweight Gloves | |
Dupont™ Kevlar™ aramid fibers | |
144 Pairs |