Learn More
4-tert-Butylstyrene, 94%, stab. with 50 ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Catalog Number p-7045887
Click to view available options
Quantity:
100 mL
500 mL
1746-23-2
C12H16
160.26
MFCD00065126
QEDJMOONZLUIMC-UHFFFAOYSA-N
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
15627
1-tert-butyl-4-ethenylbenzene
CC(C)(C)C1=CC=C(C=C1)C=C
1746-23-2
C12H16
160.26
MFCD00065126
QEDJMOONZLUIMC-UHFFFAOYSA-N
4-tert-butylstyrene, p-tert-butylstyrene, p-t-butylstyrene, benzene, 1-1,1-dimethylethyl-4-ethenyl, styrene, p-tert-butyl, unii-9pct9cbw0v, 1-1,1-dimethylethyl-4-ethenylbenzene, 9pct9cbw0v, 1-tert-butyl-4-vinylbenzene, 4-t-butyl styrene
15627
1-tert-butyl-4-ethenylbenzene
CC(C)(C)C1=CC=C(C=C1)C=C
4-tert-Butylstyrene is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-tert-Butylstyrene is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store in the dark. Protect from heat. Incompatible with heat, light, free radical initiators, UV radiation. Do not store together.
TRUSTED_SUSTAINABILITY
Melting Point -38°C
Density 0.884
Boiling Point 219°C
Flash Point 80°C (176°F)
Refractive Index 1.525
Quantity 100 mL
Beilstein 1903714
Solubility Information Not miscible in water.
Formula Weight 160.26
Percent Purity 94%
Chemical Name or Material 4-tert-Butylstyrene, stabilized with 50ppm 4-tert-butylcatechol