Learn More
4'-Acetoxyacetophenone, 99%, Thermo Scientific Chemicals
Product Code. 11445956
Click to view available options
Quantity:
25 g
100 g
500 g
4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
Solubility
Slightly soluble in water(4450 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13031-43-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017229
InChI Key SMIOEQSLJNNKQF-UHFFFAOYSA-N
Synonym 4'-acetoxyacetophenone, p-acetoxyacetophenone, 4-acetoxyacetophenone, p-acetylphenyl acetate, 4-acetylphenyl acetate, unii-eo1xk390jf, ethanone, 1-4-acetyloxy phenyl, 4-acetoxy acetophenone, 4-acetoxy-acetophenone, eo1xk390jf
PubChem CID 83063
ChEBI CHEBI:86558
IUPAC Name (4-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C(C)=O
Specifications
Melting Point 51°C to 54°C
Boiling Point 165°C to 170°C (14 mmHg)
Quantity 500 g
Beilstein 2093080
Solubility Information Slightly soluble in water(4450mg/L).
Formula Weight 178.19
Percent Purity 99%
Chemical Name or Material 4'-Acetoxyacetophenone