Learn More
(±)-2-Pentanol, 99%, Thermo Scientific Chemicals
Catalog Number p-7025624
Click to view available options
Quantity:
500 mL
2500 mL
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O
6032-29-7
C5H12O
88.15
MFCD00004579
JYVLIDXNZAXMDK-UHFFFAOYSA-N
2-pentanol, sec-amyl alcohol, methylpropylcarbinol, sec-pentyl alcohol, pentanol-2, methyl propyl carbinol, 2-pentyl alcohol, 2-hydroxypentane, 1-methyl-1-butanol, alpha-methylbutanol
22386
CHEBI:77518
pentan-2-ol
CCCC(C)O
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.
Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point °C to 50°C
Density 0.811
Boiling Point 114°C to 115°C
Flash Point 33°C (91°F)
Assay Percent Range 0.99
Refractive Index 1.406
Quantity 500 mL
UN Number UN1105
Beilstein 1718819
Merck Index 14,7119
Solubility Information Soluble in water at 20°C,166g/L. Soluble in alcohol.
Formula Weight 88.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (±)-2-Pentanol