Learn More
2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals
Catalog Number p-3758493
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
5271-38-5
C3H8OS
92.16
MFCD00002908
WBBPRCNXBQTYLF-UHFFFAOYSA-N
2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
78925
CHEBI:63861
2-methylsulfanylethanol
CSCCO
5271-38-5
C3H8OS
92.16
MFCD00002908
WBBPRCNXBQTYLF-UHFFFAOYSA-N
2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
78925
CHEBI:63861
2-methylsulfanylethanol
CSCCO
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0600g/mL
Boiling Point 169°C to 171°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3SCH2CH2OH
Refractive Index 1.4934 to 1.4954
Quantity 5 mL
Beilstein 01, II, 524
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble
Formula Weight 92.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Methylthio)ethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement