Learn More
Thermo Scientific Chemicals 2'-Fluoro-2'-deoxyuridine, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2'-Fluoro-2'-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
Solubility
Soluble in methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
2′-Fluoro-2′-deoxyuridine is used as a component incorporated into therapeutic oligonucleotides to combat prostate and breast cancer.
Solubility
Soluble in methanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Quantity | 1 g |
Solubility Information | Soluble in methanol. |
Formula Weight | 246.19 |
Percent Purity | 99% |
Physical Form | Powder |
Chemical Name or Material | 2'-Fluoro-2'-deoxyuridine |