missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Chlorophenoxy)ethanethioamide, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95%+ |
Packaging | Amber Glass Bottle |
Linear Formula | ClC6H4OCH2CSNH2 |
Quantity | 1 g |
Formula Weight | 201.68 |
Percent Purity | 95% |
Chemical Name or Material | 2-(3-Chlorophenoxy)ethanethioamide |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-(3-Chlorophenoxy)ethanethioamide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'