missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanethiol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
1-Butanethiol is used as a model gaseous analyte for fast-responding membrane-free amperometric gas sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -116°C |
Color | Colorless |
Density | 0.842 g/mL |
Boiling Point | 98°C |
Flash Point | 2°C (36°F) |
Odor | Stench |
Linear Formula | CH3(CH2)3SH |
Refractive Index | 1.4425 |
Quantity | 25 mL |
UN Number | UN2347 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'