Learn More
SPEX Certiprep 1.5GR Phthalates in Polyvinyl Chloride, 1,000 µg/mL
Brand:  SPEX Certiprep CRM-PVC003
1.5GR Phthalates in Polyvinyl Chloride, 1,000 µg/m
Product Code. 16379087
  • 136.00 EUR / 1.50g
Explore more special offers
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Phthalates in PVC
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : polyvinyl chloride,diisobutyl phthalate, di-n-pentyl phthalate, di-n-hexyl phthalate, dicyclohexyl