Learn More
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
Description
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is used as a catalyst for isomerization of allylic ethers of glycerol and glucose to the corresponding 1-propenyl ethers. It is also used as a catalyst in hydrogenation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 224°C (decomposition) |
Quantity | 0.5 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in acetone; Insoluble in water. |
Formula Weight | 845.79 |
Physical Form | Crystalline |
Chemical Name or Material | 1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate |