missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyl-1,3-divinyldisilazane, 96%, Thermo Scientific™

Product Code. 11349185
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,1,3,3-Tetramethyl-1,3-divinyldisilazane is used for silicone rubber, silicone resin, silicone gel and vinyl manufacturing.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7691-02-3
MDL Number MFCD00042846
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene

Specifications

Boiling Point 160°C to 161°C
Quantity 25 g
Molecular Formula C8H19NSi2
UN Number UN2924
Beilstein 2411661
Flash Point 42°C (107°F)
Synonym 1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
Solubility Information Hydrolyzes in water.
InChI Key WYUIWUCVZCRTRH-UHFFFAOYSA-N
SMILES C[Si](C)(C=C)N[Si](C)(C)C=C
Molecular Weight (g/mol) 185.42
PubChem CID 82126
Formula Weight 185.42
Percent Purity 96%
Refractive Index 1.44
Sensitivity Moisture sensitive
Density 0.819
Chemical Name or Material 1,1,3,3-Tetramethyl-1,3-divinyldisilazane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.