Fisherbrand Product of the Month
Fisherbrand™ Isotemp™ CO2 Incubator, 150 L, Stainless Steel
- Efficient high performance heaters surround the chamber for even temperature distribution
- Gentle fan assisted air flow pattern is designed for uniform distribution of temperature, CO2, and humidity
- The 90°C high temperature decontamination cycle simplifies cleaning because no autoclave and toxic chemicals required
- The cycle eliminates variability in disinfection and has been proven effective against wide range of contaminants including bacteria, molds, and fungal spores and mycoplasma, eliminating the variability in disinfection
- CO2 sensors are positioned directly in the chamber for precise control. A choice of CO2 sensor technology includes.
- Thermal Conductivity (TC) for precise monitoring and reliable long life
- Infrared (IR) for independent operation for precise, remote monitoring applications
Terms & Conditions
*Discount will apply to qualifying orders received by the Fisher Scientific channel between 01 June and 30 June 2025. Promotion code must be presented at the time of order. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or by local laws or regulations or agency/institutional policy. Other restrictions may apply.