missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7059519
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
23519-77-9
C12H28O4Zr
327.58
MFCD00015307
XPGAWFIWCWKDDL-UHFFFAOYSA-N
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
90139
propan-1-olate;zirconium(4+)
CCCO[Zr](OCCC)(OCCC)OCCC
This item is not returnable. View return policy
23519-77-9
C12H28O4Zr
327.58
MFCD00015307
XPGAWFIWCWKDDL-UHFFFAOYSA-N
zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
90139
propan-1-olate;zirconium(4+)
CCCO[Zr](OCCC)(OCCC)OCCC

Zirconium(IV) n-propoxide is used as a precursor for the preparation of Zirconia. It is used as coupling agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Density 1.044 g/mL
Flash Point 28°C (83°F)
Packaging ChemSeal™ bottles
Refractive Index 1.451
Quantity 50 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 327.56
Concentration or Composition (by Analyte or Components) 70% w/w in n-propanol
Physical Form Liquid
Chemical Name or Material Zirconium (IV) n-propoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'