missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, Histological Grade, Honeywell™
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Description
- Commonly used for tissue sampling and processing
- Precise control of the manufacturing process ensures unsurpassed lot-to-lot consistency
- Available in a variety of packaging options (amber glass bottles, carbon steel flex-spout cans and carbon steel cans with NPT threads)
Specifications
Specifications
Color | Colorless |
Density | 0.86g/mL at 25°C |
Boiling Point | 140°C |
Quantity | 20 L |
Explosion Limit | 0.07 |
Flash Point | 25°C (77°F) |
Packaging | Stainless Steel Can |
Physical Form | Liquid |
Refractive Index | n20/D 1.497 |
Linear Formula | C6H4(CH3)2 |
Show More |
Safety and Handling
Product Identifier
- Xylenes
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H312 + H332-Harmful in contact with skin or if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'