Learn More
Xylem Analytics™ WTW™ Type-800 Combined Ion-Selective Electrodes (ISEs)
WTW™ Type-800 Combined Ion-Selective Electrodes (ISEs) for measurement of the electrochemical concentration of selected ions. Easy to handle, compact, combined electrodes allow the measurement of small sample volumes. A cost-effective option for analyzing small amounts of material.
Brand: Xylem Analytics™ 106651
Additional Details : Weight : 0.50000kg
Description
- Product Type: Type-800 Combined Electrodes
- Use: Measurement of Electrochemical Ion Concentrations
- Connector: DIN or BNC
- Cable Length: 1 m (3.3 ft.)
Specifications
DIN | |
1 m (3.3 ft.) | |
Ag/S 800 DIN | |
Combined ion-Selective Electrode (ISE) | |
0.003 to 32,000 mg/L |
Requirements: ELY/BR/503 reference electrolyte, ionic strength adapter ISA/FK (Ag-ions) or according to instruction manual for sulphide measurement. Standard solutions have to be prepared freshly. | |
2 to 12 | |
Combined, Silver Ion/Sulfide-Selective | |
Electrode |