Learn More
DWK Life Sciences Wheaton™ Stainless Steel Support Screens
For vacuum filtration assembly. Wheaton™ Stainless Steel Support Screens are manufactured from the highest quality materials.
Brand:  DWK Life Sciences 419364
160.55 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.40830kg
 View more versions of this product
Product Code. 11591315
  • 169.00 EUR / Each
Estimated Shipment: 24-07-2024
Description
For vacuum filtration assembly. Wheaton™ Stainless Steel Support Screens are manufactured from the highest quality materials.
  • Stainless Steel
  • For use with 47mm assembly
Specifications
Specifications
47mm Vacuum Filtration Assembly