Learn More
Cytiva Whatman™ Grade 598 Cellulose Qualitative Filter Paper
A thick filter paper with high loading capacity. Combines medium retention with medium-fast to fast filtration speed.
12.30 EUR - 725.00 EUR
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
16032062
|
Cytiva
10312000 |
Roll |
570.00 EUR
1 roll |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
11393055
|
Cytiva
10312209 |
Disc |
12.30 EUR
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
11340544
|
Cytiva
10312287 |
Sheet |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Specifications
140 g/m2 | |
10 μm | |
Grade 598 |