Learn More
Macherey-Nagel™ Visocolor™ HE total Hardness High Sensitive Titration Kit
Brand:  Macherey-Nagel™ 915005
Additional Details : Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 10579101
  • 54.40 EUR / Each
Estimated Shipment: 05-06-2024
Specifications
1 Each
10 to 360mg/L CaCO3
100 Tests with an average hardness of 10 °d
HE total Hardness High Sensitive Titration Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR HE Total Hardness H 20 F
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : triethanolamine, ethanol