Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, Quick-Disconnect
VapLock™ Adapters for Exhaust Filters, Quick-Disconnect feature various adapter configurations allowing quick and easy connection to emission sources
POA
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | VapLock Exhaust Filters |
Thread Size (English) | 1/4 in. NPT(M) |
Description
- Fit VapLock exhaust filters, which have 1/4 in NPT(F) ports
- Features Justrite Centura quick disconnect
Warranty
None
Specifications
Solvent Safety | |
VapLock Exhaust Filters | |
1/4 in. NPT(M) |