Learn More
Cole-Parmer™ VapLock™ Adapters for Exhaust Filters, NPT(M)
VapLock™ Adapters for Exhaust Filters, NPT(M) feature various adapter configurations allowing quick and easy connection to emission sources
POA
Specifications
Type | Filter Adapter; 1/ea |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Exhaust Filters |
Description
- Fit VapLock exhaust filters, which have 1/4-inch NPT(F) ports
- May require additional adapters to fit your container
Warranty
None
Specifications
Filter Adapter; 1/ea | |
Solvent Safety | |
VapLock Exhaust Filters |